tert-Butyl phenyl carbonate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Carbonic Acid tert-Butyl Phenyl Ester

H60867-14 H60867-22
CAAAH60867-14EA 124.98 CAD
CAAAH60867-14 CAAAH60867-22
tert-Butyl phenyl carbonate 97%
tert-Butyl phenyl carbonate

Purity: 98% CAS No.: 6627-89-0 Molecular Formula: C11H14O3 Formula Weight: 194.23 Flash Point: 101°(214°F) Boiling Point: 83° Beilstein Reference: 2361602 MDL No.: MFCD00008804

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Boiling Pt: 83 °C
Density: 1.05
Flash Pt: 101 °C(214 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008804
CAS Number: 6627-89-0
EINECS: 229-601-8

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR