(2R,5R)-1-(2-[(2R,5R)-2,5-Dimethylphospholan-1-yl]phenyl)-2,5-dimethylphospholane-1-oxide ≥97%
Supplier: Thermo Scientific Chemicals
H60834-03
H60834-MD
CAAAH60834-03EA
684.14
CAD
CAAAH60834-03
CAAAH60834-MD
(2R,5R)-1-(2-[(2R,5R)-2,5-Dimethylphospholan-1-yl]phenyl)-2,5-dimethylphospholane-1-oxide ≥97%
(2R,5R)-1-(2-[(2R,5R)-2,5-Dimethylphospholan-1-yl]phenyl)-2,5-dimethylphospholane-1-oxide
Purity: 97+%
CAS No.: 638132-66-8
Molecular Formula: C18H28OP2
Formula Weight: 322.36
Melting Point: 116-122°
Optical Rotation: -171° (c=1 in chloroform)
MDL No.: MFCD09750450
Formula:
C₁₈H₂₈OP₂ Melting Pt: 116…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD09750450 CAS Number: 638132-66-8 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...