4-Vinyl-1,3-dioxolan-2-one 99%

Supplier: Thermo Scientific Chemicals
Danger

H60822-18 H60822-30
CAAAH60822-18EA 128.77 CAD
CAAAH60822-18 CAAAH60822-30
4-Vinyl-1,3-dioxolan-2-one 99%
4-Vinyl-1,3-dioxolan-2-one

Purity: 99% CAS No.: 4427-96-7 Molecular Formula: C5H6O3 Formula Weight: 114.10 Storage Temperature: 2°C to 8°C Flash Point: 96°(205°F) Refractive Index: 1.45 Boiling Point: 237° UN No.: 2810 MDL No.: MFCD00143315

Formula: C₅H₆O₃
MW: 114.10 g/mol
Boiling Pt: 237 °C
Density: 1.188
Flash Pt: 96 °C(205 °F)
Storage Temperature: Ambient
MDL Number: MFCD00143315
CAS Number: 4427-96-7
UN: 2810
ADR: "6.1",III

Order Now

Specification Test Results

Assay Minimum 99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR