2-Phenylthiophene 95%

Supplier: Thermo Scientific Chemicals
Warning

H60819-03 H60819-06
CAAAH60819-03EA 55.14 CAD
CAAAH60819-03 CAAAH60819-06
2-Phenylthiophene 95%
2-Phenylthiophene

CAS: 825-55-8 MDL: MFCD00130080

Formula: C₁₀H₈S
MW: 160.24 g/mol
Boiling Pt: 256 °C
Melting Pt: 34…36 °C
Flash Pt: 109 °C(228 °F)
Storage Temperature: Ambient
MDL Number: MFCD00130080
CAS Number: 825-55-8

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR