Piperonyl butoxide tech. 90%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
H60771-AC
H60771-AE
CAAAH60771-ACEA
36.5
CAD
CAAAH60771-AC
CAAAH60771-AE
Piperonyl butoxide tech. 90%, Technical Grade
Piperonyl butoxide
Piperonyl butoxide is used as a pharmaceutical intermediate.
Purity: 90%
CAS No.: 51-03-6
Molecular Formula: C19H30O5
Formula Weight: 338.44
Flash Point: 171°(340°F)
Refractive Index: 1.4980
Boiling Point: 155°/0.3mm
Beilstein Reference: 288063
UN No.: 3082
MDL No.: MFCD00005842
Soluble in benzene, ethanol and methanol
Formula:
C₁₉H₃₀O₅ Boiling Pt: 155 °C/0.3 mm Density: 1.059 Flash Pt: 171 °C(340 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005842 CAS Number: 51-03-6 EINECS: 200-076-7 UN: 3082 ADR: "9",III |
Specification Test Results
Assay Minimum | 90 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...