(5R,6S)-(-)-4-Boc-5,6-diphenyl-2-morpholinone 98%

Supplier: Thermo Scientific Chemicals
Warning

H60764-03 H60764-06
CAAAH60764-03EA 127.82 CAD
CAAAH60764-03 CAAAH60764-06
(5R,6S)-(-)-4-Boc-5,6-diphenyl-2-morpholinone 98%
(5R,6S)-(-)-4-Boc-5,6-diphenyl-2-morpholinone

Purity: 98% CAS No.: 112741-49-8 Molecular Formula: C21H23NO4 Formula Weight: 353.41 Storage Temperature: 2°C to 8°C Physical Form: Solid Melting Point: 206° dec. Optical Rotation: -87° (c=5.5 in methylene chloride) MDL No.: MFCD00074955

Formula: C₂₁H₂₃NO₄
Melting Pt: 206 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00074955
CAS Number: 112741-49-8

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR