2-[4-(Bromomethyl)phenyl]propionic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Bromomethyl)hydratropic Acid, 2-(4-(Bromomethyl)phenyl)propanoic acid

H60763-06 H60763-14
CAAAH60763-06EA 132.88 CAD
CAAAH60763-06 CAAAH60763-14
2-[4-(Bromomethyl)phenyl]propionic acid 96%
2-[4-(Bromomethyl)phenyl]propionic acid

Purity: 96% CAS No.: 111128-12-2 Molecular Formula: C10H11BrO2 Formula Weight: 243.10 Melting Point: 126-130° UN No.: 3261 MDL No.: MFCD02093445

Formula: C₁₀H₁₁BrO₂
Melting Pt: 126…130 °C
Storage Temperature: Ambient
MDL Number: MFCD02093445
CAS Number: 111128-12-2
ADR: 8,III

Order Now

Specification Test Results

Assay Minimum 96


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR