1-Propanesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Propane-1-sulphonyl chloride, Propane-1-sulfonyl chloride

H60747-AC H60747-AE
CAAAH60747-ACEA 76.79 CAD
CAAAH60747-AC CAAAH60747-AE
1-Propanesulfonyl chloride 97%
1-Propanesulphonyl chloride

CAS: 10147-36-1 MDL: MFCD00007462

Formula: C₃H₇ClO₂S
MW: 142.61 g/mol
Boiling Pt: 78…79 °C
Melting Pt: –46 °C
Density: 1.267
Flash Pt: 97 °C (207 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007462
CAS Number: 10147-36-1
EINECS: 233-414-7
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR