10-Hydroxydecanoic acid 96%
Supplier: Thermo Scientific Chemicals
H60743-03
H60743-06
CAAAH60743-03EA
141.88
CAD
CAAAH60743-03
CAAAH60743-06
10-Hydroxydecanoic acid 96%
10-Hydroxydecanoic acid
CAS: 1679-53-4
MDL: MFCD00010510
Formula:
C₁₀H₂₀O₃ Melting Pt: 75…77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010510 CAS Number: 1679-53-4 EINECS: 216-848-1 |
Specification Test Results
Assay Minimum | 96 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...