3,4,6-Tri-O-methyl-D-glucal 97%

Supplier: Thermo Scientific Chemicals

H60694-03 H60694-MD
CAAAH60694-03EA 222.78 CAD
CAAAH60694-03 CAAAH60694-MD
3,4,6-Tri-O-methyl-D-glucal 97%
3,4,6-Tri-O-methyl-D-glucal

Purity: 97% CAS No.: 16740-98-0 Molecular Formula: C9H16O4 Formula Weight: 188.22 Storage Temperature: 2°C to 8°C MDL No.: MFCD22989010

Formula: C₉H₁₆O₄
Storage Temperature: Ambient
MDL Number: MFCD22989010
CAS Number: 16740-98-0

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR