Trimethylstearylammonium chloride 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trimethylstearylammonium Chloride

H60665-04 H60665-09
CAAAH60665-04EA 65.41 CAD
CAAAH60665-04 CAAAH60665-09
Trimethylstearylammonium chloride 95%
Trimethylstearylammonium chloride

Purity: 95% CAS No.: 112-03-8 Molecular Formula: C21H46NCl Formula Weight: 348.05 Beilstein Reference: 3917847 MDL No.: MFCD00050188

Formula: C₂₁H₄₆ClN
Storage Temperature: Ambient
MDL Number: MFCD00050188
CAS Number: 112-03-8
EINECS: 203-929-1
UN: 3261
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR