1H-Pyrazole-1-carboximidamide hydrochloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H60631-09
H60631-18
CAAAH60631-09EA
91.8
CAD
CAAAH60631-09
CAAAH60631-18
1H-Pyrazole-1-carboximidamide hydrochloride 99%
1H-Pyrazole-1-carboximidamide hydrochloride
Purity: 99%
CAS No.: 4023-02-3
Molecular Formula: C4H6N4.HCl
Formula Weight: 146.58
Melting Point: 167-170°
MDL No.: MFCD00210087
Formula:
C4H7ClN4 Melting Pt: 167…170 °C Storage Temperature: Ambient |
MDL Number:
MFCD00210087 CAS Number: 4023-02-3 EINECS: 429-520-1 |
Specification Test Results
Assay Minimum | 99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...