(R)-Methyl-2-amino-3-(1H-indol-3-yl)propanoate hydrochloride 98%

Supplier: Thermo Scientific Chemicals

Synonyms: H-D-Trp-OMe·HCl, D-Tryptophan methyl ester hydrochloride, H-D-Trp-OMe.HCl

H60622-03 H60622-06
CAAAH60622-03EA 42.34 CAD
CAAAH60622-03 CAAAH60622-06
(R)-Methyl-2-amino-3-(1H-indol-3-yl)propanoate hydrochloride 98%
(R)-Methyl-2-amino-3-(1H-indol-3-yl)propanoate hydrochloride

Purity: 98% CAS No.: 14907-27-8 Molecular Formula: C12H14N2O2.HCl Formula Weight: 254.71 Melting Point: 213-216° Optical Rotation: -18° (c=5 in methanol) MDL No.: MFCD00038992

Formula: C₁₂H₁₄N₂O₂·HCl
Melting Pt: 213…216 °C
Storage Temperature: Ambient
MDL Number: MFCD00038992
CAS Number: 14907-27-8

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR