Diethyl iminodiacetate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H60608-AA
H60608-AC
CAAAH60608-AAEA
53.88
CAD
CAAAH60608-AA
CAAAH60608-AC
Diethyl iminodiacetate 97%
Diethyl iminodiacetate
Purity: 97%
CAS No.: 6290-05-7
Molecular Formula: C8H15NO4
Formula Weight: 189.21
Flash Point: 113°(235°F)
Refractive Index: 1.435
Boiling Point: 208°
MDL No.: MFCD00041925
Formula:
C₈H₁₅NO₄ Boiling Pt: 208 °C Density: 1.056 Flash Pt: 113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041925 CAS Number: 6290-05-7 EINECS: 228-533-6 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...