5-Bromo-2,4-dimethoxybenzoic acid 97%

Supplier: Thermo Scientific Chemicals

H60599-03 H60599-06
CAAAH60599-03EA 86.58 CAD
CAAAH60599-03 CAAAH60599-06
5-Bromo-2,4-dimethoxybenzoic acid 97%
5-Bromo-2,4-dimethoxybenzoic acid

Purity: 97% CAS No.: 32246-20-1 Molecular Formula: C9H9BrO4 Formula Weight: 261.07 Melting Point: 198-202° MDL No.: MFCD05865172

Formula: C₉H₉BrO₄
Melting Pt: 198…202 °C
Storage Temperature: Ambient
MDL Number: MFCD05865172
CAS Number: 32246-20-1

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR