1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene ≥97%

Supplier: Thermo Scientific Chemicals

Synonyms: (-)-1,2-Bis((2R,5R)-2,5-dimethylphospholano)benzene, 1,2-Bis((2R,5R)-2,5-dimethylphospholan-1-yl)benzene, 1,2-Bis[(2R,5R)-2,5-dimethylphospholan-1-yl]benzene, 1,2-Bis[(2R,5R)-2,5-dimethylphospholano]benzene, (R,R)-Me-DUPHOS

H60592-03 H60592-MD
CAAAH60592-03EA 736.28 CAD
CAAAH60592-03 CAAAH60592-MD
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene ≥97%
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene

Purity: 97+% CAS No.: 147253-67-6 Molecular Formula: C18H28P2 Formula Weight: 306.36 Melting Point: 66-76° Beilstein Reference: 4810602 MDL No.: MFCD00142336

Formula: C₁₈H₂₈P₂
Melting Pt: 66…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00142336
CAS Number: 147253-67-6

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR