1,1,1,3,5,5,5-Heptamethyltrisiloxane 97%

Supplier: Thermo Scientific Chemicals
Danger

H60586-AD H60586-AK
CAAAH60586-ADEA 175.38 CAD
CAAAH60586-AD CAAAH60586-AK
1,1,1,3,5,5,5-Heptamethyltrisiloxane 97%
1,1,1,3,5,5,5-Heptamethyltrisiloxane

CAS: 1873-88-7 MDL: MFCD00048000

Formula: C₇H₂₂O₂Si₃
Boiling Pt: 142 °C
Density: 0.819
Flash Pt: 22 °C (72 °F)
Storage Temperature: Ambient
MDL Number: MFCD00048000
CAS Number: 1873-88-7
EINECS: 217-496-1
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR