4-(4-Biphenylyl)-4-oxobutyric acid 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Fenbufen, 4-([1,1'-Biphenyl]-4-yl)-4-oxobutanoic acid, Cinopal, Napanol, Lederfen, Bufemid

H60557-03 H60557-06
CAAAH60557-03EA 88.8 CAD
CAAAH60557-03 CAAAH60557-06
4-(4-Biphenylyl)-4-oxobutyric acid 96%
4-(4-Biphenylyl)-4-oxobutyric acid

Purity: 96% CAS No.: 36330-85-5 Molecular Formula: C16H14O3 Formula Weight: 254.28 Melting Point: 184-187° UN No.: 2811 MDL No.: MFCD00056701

Formula: C₁₆H₁₄O₃
MW: 254.285 g/mol
Melting Pt: 184…187 °C
Storage Temperature: Ambient
MDL Number: MFCD00056701
CAS Number: 36330-85-5
EINECS: 252-979-0
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Assay Minimum 96


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR