(R)-(-)-8-Diphenylphosphino-1,2,3,4-tetrahydro-1-naphthylamine ≥97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-1-Amino-8-(diphenylphosphino)-1,2,3,4-tetrahydronaphthalene, (R)-8-(Diphenylphosphino)-1,2,3,4-tetrahydronaphthalen-1-amine

H60538-03 H60538-MD
CAAAH60538-03EA 609.88 CAD
CAAAH60538-03 CAAAH60538-MD
(R)-(-)-8-Diphenylphosphino-1,2,3,4-tetrahydro-1-naphthylamine ≥97%
(R)-(-)-8-Diphenylphosphino-1,2,3,4-tetrahydro-1-naphthylamine

Purity: 97+% CAS No.: 960128-64-7 Molecular Formula: C22H22NP Formula Weight: 331.39 Melting Point: 120-127° Optical Rotation: -60° (c=0.5 in chloroform) MDL No.: MFCD16621442

Formula: C₂₂H₂₂NP
MW: 331.399 g/mol
Melting Pt: 120…127 °C
Storage Temperature: Ambient
MDL Number: MFCD16621442
CAS Number: 960128-64-7
UN: 3335
ADR: 9 III

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR