(S)-(+)-1-Benzyloxy-2-propanol 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S)-1-Benzyloxy-2-propanol, Benzyl (S)-(+)-2-Hydroxypropyl Ether
H60523-03
H60523-06
CAAAH60523-03EA
82.48
CAD
CAAAH60523-03
CAAAH60523-06
(S)-(+)-1-Benzyloxy-2-propanol 96%
(S)-(+)-1-Benzyloxy-2-propanol
Purity: 96%
CAS No.: 85483-97-2
Molecular Formula: C10H14O2
Formula Weight: 166.22
Flash Point: 108°(226°F)
Refractive Index: 1.51
Optical Rotation: +14° (c=1 in chloroform)
MDL No.: MFCD07367008
Formula:
C₁₀H₁₄O₂ MW: 166.22 g/mol Density: 1.044 Flash Pt: 108 °C(226 °F) Storage Temperature: Ambient |
MDL Number:
MFCD07367008 CAS Number: 85483-97-2 |
Specification Test Results
Assay Minimum | 96 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...