3,5-Dimethoxybenzylamine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H60503-06
H60503-14
CAAAH60503-06EA
169.06
CAD
CAAAH60503-06
CAAAH60503-14
3,5-Dimethoxybenzylamine 98%
3,5-Dimethoxybenzylamine
CAS: 34967-24-3
MDL: MFCD00052813
Formula:
C₉H₁₃NO₂ Boiling Pt: 94…96 °C Melting Pt: 35…37 °C Density: 1.106 Flash Pt: 113 °C (235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00052813 CAS Number: 34967-24-3 UN: 3259 ADR: 8,III |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...