1,2-Benzisothiazol-3(2H)-one 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H60492-06
H60492-14
CAAAH60492-06EA
138.57
CAD
CAAAH60492-06
CAAAH60492-14
1,2-Benzisothiazol-3(2H)-one 97%
1,2-Benzisothiazol-3(2H)-one
CAS: 2634-33-5
MDL: MFCD00127753
Formula:
C₇H₅NOS Melting Pt: 154…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044001 CAS Number: 2634-33-5 EINECS: 220-120-9 UN: 3077 ADR: 9,III |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...