3-(2-Aminoethyl)-1-boc-azetidine 95%

Supplier: Thermo Scientific Chemicals
Danger

H60469-03 H60469-MD
CAAAH60469-03EA 741.02 CAD
CAAAH60469-03 CAAAH60469-MD
3-(2-Aminoethyl)-1-boc-azetidine 95%
3-(2-Aminoethyl)-1-boc-azetidine

Purity: 95% CAS No.: 898271-20-0 Molecular Formula: C10H20N2O2 Formula Weight: 200.28 MDL No.: MFCD09878606

Formula: C₁₀H₂₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09878606
CAS Number: 898271-20-0
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR