Methyl (±)-lactate 99%

Supplier: Thermo Scientific Chemicals
Warning

H60455-AA H60455-AC
CAAAH60455-AAEA 109.97 CAD
CAAAH60455-AA CAAAH60455-AC
Methyl (±)-lactate 99%
Methyl (±)-lactate

CAS: 547-64-8 MDL: MFCD00066367

Formula: C₄H₈O₃
Boiling Pt: 35 °C (1013 hPa)
Density: 1.093
Flash Pt: 51 °C (124 °F)
Storage Temperature: Ambient
MDL Number: MFCD00066367
CAS Number: 547-64-8
EINECS: 208-930-0
UN: 3272
ADR: 3,III
Merck Index: 12,06167

Order Now

Specification Test Results

Assay Minimum 99


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR