N-(3-Hydroxypropyl)morpholine 95%

Supplier: Thermo Scientific Chemicals
Warning

H60376-03 H60376-06
CAAAH60376-03EA 67.31 CAD
CAAAH60376-03 CAAAH60376-06
N-(3-Hydroxypropyl)morpholine 95%
N-(3-Hydroxypropyl)morpholine

CAS: 4441-30-9 MDL: MFCD01713533

Formula: C₇H₁₅NO₂
Boiling Pt: 240…241 °C
Density: 1.049
Flash Pt: 85 °C(185 °F)
Storage Temperature: Ambient
MDL Number: MFCD01713533
CAS Number: 4441-30-9

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR