1-(2-Hydroxyethyl)pyrrolidine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H60368-AC
H60368-AE
CAAAH60368-ACEA
85.48
CAD
CAAAH60368-AC
CAAAH60368-AE
1-(2-Hydroxyethyl)pyrrolidine 97%
1-(2-Hydroxyethyl)pyrrolidine
Purity: 97%
CAS No.: 2955-88-6
Molecular Formula: C6H13NO
Formula Weight: 115.17
Flash Point: 56°(133°F)
Refractive Index: 1.471
Boiling Point: 79-81°
Beilstein Reference: 102976
UN No.: 1993
MDL No.: MFCD00003181
Formula:
C₆H₁₃NO Boiling Pt: 79…81 °C Density: 0.985 Flash Pt: 56 °C(133 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003181 CAS Number: 2955-88-6 EINECS: 220-976-3 UN: 1993 ADR: "3",III |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...