1,5-Diphenylcarbazone ≥95%

Supplier: Thermo Scientific Chemicals

H60336-06 H60336-14
CAAAH60336-06EA 135.88 CAD
CAAAH60336-06 CAAAH60336-14
1,5-Diphenylcarbazone ≥95%
1,5-Diphenylcarbazone

CAS: 538-62-5 MDL: MFCD00003024

Formula: C₁₃H₁₂N₄O
Boiling Pt: 440 °C (1013 hPa)
Melting Pt: 119…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00003024
CAS Number: 538-62-5
EINECS: 208-698-0
Merck Index: 12,03380

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR