4-Bromo-N,N-diphenylaniline 99%

Supplier: Thermo Scientific Chemicals
Warning

H60328-06 H60328-14
CAAAH60328-06EA 133.67 CAD
CAAAH60328-06 CAAAH60328-14
4-Bromo-N,N-diphenylaniline 99%
4-Bromo-N,N-diphenylaniline

CAS: 36809-26-4 MDL: MFCD01851266

Formula: C₁₈H₁₄BrN
Melting Pt: 108…112 °C
Storage Temperature: Ambient
MDL Number: MFCD01851266
CAS Number: 36809-26-4

Order Now

Specification Test Results

Assay Minimum 99


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR