Bis[2-(dicyclohexylphosphino)ethyl]amine ≥97%

Supplier: Thermo Scientific Chemicals
Danger

H60269-03 H60269-06 H60269-MD
CAAAH60269-03EA 262.28 CAD
CAAAH60269-03 CAAAH60269-06 CAAAH60269-MD
Bis[2-(dicyclohexylphosphino)ethyl]amine ≥97%
Bis[2-(dicyclohexylphosphino)ethyl]amine

Purity: 97+% CAS No.: 550373-32-5 Molecular Formula: C28H53NP2 Formula Weight: 465.67 Melting Point: 64-69° UN No.: 3259 MDL No.: MFCD17014019

Formula: C₂₈H₅₃NP₂
Melting Pt: 64…69 °C
Storage Temperature: Ambient
MDL Number: MFCD17014019
CAS Number: 550373-32-5
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR