2-Methyl-5-nitroimidazole-1-ethanol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H60258-14
H60258-22
CAAAH60258-14EA
158
CAD
CAAAH60258-14
CAAAH60258-22
2-Methyl-5-nitroimidazole-1-ethanol 99%
2-Methyl-5-nitroimidazole-1-ethanol
Purity: 99%
CAS No.: 443-48-1
Molecular Formula: C6H9N3O3
Formula Weight: 171.16
Storage Temperature: 2°C to 8°C
Melting Point: 159-161°
Beilstein Reference: 611683
MDL No.: MFCD00009750
Formula:
C₆H₉N₃O₃ Melting Pt: 159…161 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009750 CAS Number: 443-48-1 EINECS: 207-136-1 |
Specification Test Results
Assay Minimum | 99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...