5-Methyl-1H-tetrazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H60248-06
H60248-14
CAAAH60248-06EA
86.9
CAD
CAAAH60248-06
CAAAH60248-14
5-Methyl-1H-tetrazole 97%
5-Methyl-1H-tetrazole
CAS: 4076-36-2
MDL: MFCD00129971
Formula:
C₂H₄N₄ MW: 84.08 g/mol Melting Pt: 142…146 °C Storage Temperature: Ambient |
MDL Number:
MFCD00185824 CAS Number: 4076-36-2 EINECS: 218-746-2 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...