3,3',5,5'-Tetramethylbenzidine dihydrochloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dihydrochloride TMB, TMB-HCl, TMB 2HCl, TMB dihydrochloride

H60245-03 H60245-MD
CAAAH60245-03EA 195.92 CAD
CAAAH60245-03 CAAAH60245-MD
3,3',5,5'-Tetramethylbenzidine dihydrochloride 99%
3,3',5,5'-Tetramethylbenzidine dihydrochloride

CAS: 64285-73-0 MDL: MFCD00012961

Formula: C₁₆H₂₂Cl₂N₂
Melting Pt: 300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012961
CAS Number: 64285-73-0
EINECS: 264-769-6

Order Now

Specification Test Results

Assay Minimum 99


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR