Methyl-2-methyl-3-nitrobenzoate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl 2-methyl-3-nitrobenzoate

H60242-06 H60242-14
CAAAH60242-06EA 60.04 CAD
CAAAH60242-06 CAAAH60242-14
Methyl-2-methyl-3-nitrobenzoate 97%
Methyl 2-methyl-3-nitrobenzoate

CAS: 59382-59-1 MDL: MFCD00082564

Formula: C₉H₉NO₄
Melting Pt: 62…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00082564
CAS Number: 59382-59-1

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR