2,4-Dibromothiazole 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,4-dibromo-1,3-thiazole

H60230-03 H60230-06
CAAAH60230-03EA 77.89 CAD
CAAAH60230-03 CAAAH60230-06
2,4-Dibromothiazole 97%
2,4-Dibromothiazole

CAS: 4175-77-3 MDL: MFCD01318994

Formula: C₃HBr₂NS
Melting Pt: 80…84 °C
Storage Temperature: Ambient
MDL Number: MFCD01318994
CAS Number: 4175-77-3

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR