6,7-Dihydrobenzo[b]thiophen-4-one 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Oxo-4,5,6,7-tetrahydrothianaphthene, 6,7-Dihydro-4-benzo[b]thiophenone, 4-Keto-4,5,6,7-tetrahydrothianaphthene
H60220-04
H60220-09
CAAAH60220-04EA
151.68
CAD
CAAAH60220-04
CAAAH60220-09
6,7-Dihydrobenzo[b]thiophen-4-one 98%
6,7-Dihydrobenzo[b]thiophen-4-one
Purity: 98%
CAS No.: 13414-95-4
Molecular Formula: C8H8OS
Formula Weight: 152.21
Flash Point: 110°(230°F)
Refractive Index: 1.5895
Melting Point: 38-40°
Boiling Point: 120°
Beilstein Reference: 112276
MDL No.: MFCD00005861
Formula:
C₈H₈OS MW: 152.211 g/mol Boiling Pt: 120 °C Melting Pt: 38…40 °C Flash Pt: 110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005861 CAS Number: 13414-95-4 EINECS: 236-510-7 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...