Benzyl chloromethyl ether ≥70%, tech.
Supplier: Thermo Scientific Chemicals
|
Danger
|
H60207-AD
H60207-AK
CAAAH60207-ADEA
202.24
CAD
CAAAH60207-AD
CAAAH60207-AK
Benzyl chloromethyl ether ≥70%, tech.
Benzyl chloromethyl ether
CAS: 3587-60-8
MDL: MFCD00000886
Formula:
C₈H₉ClO Boiling Pt: 102 °C Density: 1.126 Storage Temperature: Ambient |
MDL Number:
MFCD00000886 CAS Number: 3587-60-8 UN: 2810 ADR: 6.1,III |
Specification Test Results
Assay Minimum | 70 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...