Benzyldimethyldodecylammonium chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Benzyldimethyldodecylammonium chloride, N-Benzyl-N,N-dimethyldodecan-1-aminium chloride, Dodecyldimethylbenzylammonium chloride, Benzyldodecyldimethylammonium chloride dihydrate

H60196-06 H60196-18
CAAAH60196-06EA 95.59 CAD
CAAAH60196-06 CAAAH60196-18
Benzyldimethyldodecylammonium chloride 98%
Benzyldimethyldodecylammonium chloride

CAS: 139-07-1 MDL: MFCD00137276

Formula: C₂₁H₃₈ClN
MW: 339.99 g/mol
Melting Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00137276
CAS Number: 139-07-1
EINECS: 205-351-5
UN: 3261
ADR: 8 III

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR