(S)-(-)-1,1,2-Triphenyl-1,2-ethanediol 98%

Supplier: Thermo Scientific Chemicals
Warning

H60189-03 H60189-06
CAAAH60189-03EA 114.08 CAD
CAAAH60189-03 CAAAH60189-06
(S)-(-)-1,1,2-Triphenyl-1,2-ethanediol 98%
(S)-(-)-1,1,2-Triphenyl-1,2-ethanediol

Purity: 98% CAS No.: 108998-83-0 Molecular Formula: C20H18O2 Formula Weight: 290.36 Physical Form: Powder Appearance: Pale brown Melting Point: 125-127° Beilstein Reference: 3208922 Optical Rotation: -214° (c=1 in ethanol) MDL No.: MFCD00075492

Formula: C₂₀H₁₈O₂
Melting Pt: 125…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00075492
CAS Number: 108998-83-0

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR