Folinic acid calcium salt, hydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

H60181-03 H60181-MD
CAAAH60181-03EA 696.78 CAD
CAAAH60181-03 CAAAH60181-MD
Folinic acid calcium salt, hydrate 98%
Folinic acid calcium salt

Purity: 98% CAS No.: 1492-18-8 Molecular Formula: C20H21CaN7O7.xH2O Formula Weight: 511.51 (anhy) Beilstein Reference: 5723924 MDL No.: MFCD00149463

Formula: C₂₀H₂₁CaN₇O₇.xH₂0
Storage Temperature: Ambient
MDL Number: MFCD00149463
CAS Number: 1492-18-8
EINECS: 216-082-8

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR