2-Piperidinemethanol 97%

Supplier: Thermo Scientific Chemicals
Warning

H60178-09 H60178-18
CAAAH60178-09EA 120.55 CAD
CAAAH60178-09 CAAAH60178-18
2-Piperidinemethanol 97%
2-Piperidinemethanol

CAS: 3433-37-2 MDL: MFCD00005987

Formula: C₆H₁₃NO
Boiling Pt: 100…102 °C
Melting Pt: 68…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00005987
CAS Number: 3433-37-2
EINECS: 222-333-2

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR