(S)-(-)-β-Citronellol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S)-(-)-3,7-Dimethyl-6-octen-1-ol, (S)-(-)-ß-Citronellol, (S)-(-)-Citronellol, (-)-beta-Citronellol, (-)-β-Citronellol, (S)-3,7-Dimethyloct-6-en-1-ol, beta-Rhodinol
H60174-03
H60174-06
CAAAH60174-03EA
65.57
CAD
CAAAH60174-03
CAAAH60174-06
(S)-(-)-β-Citronellol 97%
(S)-(-)-β-Citronellol
CAS: 7540-51-4
MDL: MFCD00063214
Formula:
C₁₀H₂₀O MW: 156.27 g/mol Boiling Pt: 225…226 °C Density: 0.856 Flash Pt: 99 °C(210 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00063214 CAS Number: 7540-51-4 EINECS: 231-415-7 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...