(1R,2R)-(+)-1,2-Cyclohexanediamine-L-tartrate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(1R,2R)-Cyclohexane-1,2-diamine (2R,3R)-2,3-dihydroxysuccinate, (1R)-trans-1,2-Diaminocyclohexane L-Tartrate
H60172-04
H60172-09
CAAAH60172-04EA
59.57
CAD
CAAAH60172-04
CAAAH60172-09
(1R,2R)-(+)-1,2-Cyclohexanediamine-L-tartrate 98%
(1R,2R)-(+)-1,2-Cyclohexanediamine-L-tartrate
(1R,2R)-(+)-1,2-Diaminocyclohexane L-tartrate is a useful chemical for the synthesis of chiral ligands.
Purity: 99%
CAS No.: 39961-95-0
Molecular Formula: C10H20N2O6
Formula Weight: 264.28
Melting Point: 273° dec.
Optical Rotation: +12° (c=4 in water)
MDL No.: MFCD00191979
Formula:
C₆H₁₄N₂ MW: 264.28 g/mol Melting Pt: 273 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00191979 CAS Number: 39961-95-0 |
Specification Test Results
Assay | ≥97.5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...