4-Methoxycinnamic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(4-Methoxyphenyl)acrylic acid

H60144-14 H60144-22
CAAAH60144-14EA 104.12 CAD
CAAAH60144-14 CAAAH60144-22
4-Methoxycinnamic acid 99%
4-Methoxycinnamic acid

Purity: 99% CAS No.: 830-09-1 Molecular Formula: C10H10O3 Formula Weight: 178.18 Melting Point: 170-173° MDL No.: MFCD00004398

Formula: C₁₀H₁₀O₃
MW: 178.18 g/mol
Melting Pt: 170…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00004398
CAS Number: 830-09-1
EINECS: 212-594-0

Order Now

Specification Test Results

Assay Minimum 99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR