kappa-Carrageenan

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Irish moss

H60117-06 H60117-14
CAAAH60117-06EA 78.05 CAD
CAAAH60117-06 CAAAH60117-14
kappa-Carrageenan
kappa-Carrageenan

CAS No.: 11114-20-8 Molecular Formula: C12H17O13S Formula Weight: 401.32 Physical Form: Solid Appearance: White MDL No.: MFCD00151514

Formula: C₁₂H₁₇O₁₃S
MW: 401.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151514
CAS Number: 11114-20-8
EINECS: 234-350-2
UN: 2811
ADR: 6.1 III
Merck Index: 14,1864

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR