1,3-Adamantanedicarboxylic acid 98%
Supplier: Thermo Scientific Chemicals
H60081-03
H60081-06
CAAAH60081-03EA
94.33
CAD
CAAAH60081-03
CAAAH60081-06
1,3-Adamantanedicarboxylic acid 98%
1,3-Adamantanedicarboxylic acid
Purity: 98%
CAS No.: 39269-10-8
Molecular Formula: C12H16O4
Formula Weight: 224.25
Melting Point: 276-278°
MDL No.: MFCD00167790
Formula:
C₁₂H₁₆O₄ Melting Pt: 276…278 °C Storage Temperature: Ambient |
MDL Number:
MFCD00167790 CAS Number: 39269-10-8 EINECS: 254-395-1 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...