Methyl-4-vinylbenzoate 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H60057-03
H60057-06
CAAAH60057-03EA
252.8
CAD
CAAAH60057-03
CAAAH60057-06
Methyl-4-vinylbenzoate 96%
Methyl-4-vinylbenzoate
Purity: 97%
CAS No.: 1076-96-6
Molecular Formula: C10H10O2
Formula Weight: 162.19
Melting Point: 32-37°
MDL No.: MFCD00017218
Formula:
C₁₀H₁₀O₂ Melting Pt: 32…37 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017218 CAS Number: 1076-96-6 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...