Ethyl-2-hydrazinylacetate hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

H60042-06 H60042-14
CAAAH60042-06EA 282.82 CAD
CAAAH60042-06 CAAAH60042-14
Ethyl-2-hydrazinylacetate hydrochloride 97%
Ethyl-2-hydrazinylacetate hydrochloride

Purity: 97% CAS No.: 6945-92-2 Molecular Formula: C4H10N2O2.HCl Formula Weight: 154.60 Melting Point: 152-154° Beilstein Reference: 3912112 MDL No.: MFCD00012923

Formula: C₄H₁₁ClN₂O₂
Melting Pt: 152…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00012923
CAS Number: 6945-92-2
EINECS: 230-104-3

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR