Iron(III) trifluoromethanesulfonate tech. 90%, tech.

Supplier: Thermo Scientific Chemicals
Warning

H60034-03 H60034-MD
CAAAH60034-03EA 86.58 CAD
CAAAH60034-03 CAAAH60034-MD
Iron(III) trifluoromethanesulfonate tech. 90%, tech.
Iron (III) trifluoromethanesulphonate

CAS: 63295-48-7 MDL: MFCD15144785

Formula: C₃F₉FeO₉S₃
Melting Pt: 183 °C
Flash Pt: 110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD15144785
CAS Number: 63295-48-7

Order Now

Specification Test Results

Assay Minimum 90

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR