1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]ethane ≥97%
Supplier: Thermo Scientific Chemicals
H60017-03
H60017-MD
CAAAH60017-03EA
807.38
CAD
CAAAH60017-03
CAAAH60017-MD
1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]ethane ≥97%
1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]ethane
Purity: 97+%
CAS No.: 528854-34-4
Molecular Formula: C22H44P2
Formula Weight: 370.53
Melting Point: 67-72°
MDL No.: MFCD08705244
Formula:
C₂₂H₄₄P₂ Melting Pt: 67…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD08705244 CAS Number: 528854-34-4 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...