1-Benzyl-3-methylimidazolium chloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Benzyl-3-methyl-1H-imidazol-3-ium chloride

H59997-06 H59997-18
CAAAH59997-06EA 58.93 CAD
CAAAH59997-06 CAAAH59997-18
1-Benzyl-3-methylimidazolium chloride 97%
1-Benzyl-3-methylimidazolium chloride

CAS: 36443-80-8 MDL: MFCD07784451

Formula: C₁₁H₁₃ClN₂
MW: 208.69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07784451
CAS Number: 36443-80-8

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR